Assay Details
Gene Symbol: LOC5569538 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652800.2 | XP_001652850.2 | 4 - 5 | 6040 | 99 | |
XM_021851080.1 | XP_021706772.1 | 4 - 5 | 6010 | 99 | ||
GenBank mRNA | AY438004.1 | - | 4 - 5 | 5989 | 99 |
