Assay Details
Gene Symbol: LOC5570276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658982.2 | XP_001659032.1 | 1 - 2 | 391 | 70 | |
GenBank mRNA | AY445078.1 | - | 1 - 2 | 355 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001658982.2 transcripts.
Set Membership: |
![]() ![]() |
