Assay Details
Gene Symbol: LOC5571497 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659697.2 | XP_001659747.2 | 6 - 7 | 4328 | 64 | |
GenBank mRNA | AY713295.1 | - | 6 - 7 | 4295 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
