Assay Details
Gene Symbol: LOC5579924 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652162.2 | XP_001652212.1 | 9 - 10 | 2936 | 83 | |
XM_021850223.1 | XP_021705915.1 | 9 - 10 | 2748 | 83 | ||
GenBank mRNA | AY713296.1 | - | 9 - 10 | 2924 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
