Assay Details
Gene Symbol: LOC5569526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652790.3 | XP_001652840.2 | 2 - 3 | 682 | 77 | |
XM_021848336.1 | XP_021704028.1 | 2 - 3 | 534 | 77 | ||
XM_021848337.1 | XP_021704029.1 | 2 - 3 | 505 | 77 | ||
GenBank mRNA | AY748242.1 | - | 2 - 3 | 475 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001652790.3, XM_021848336.1, XM_021848337.1 transcripts.
Set Membership: |
![]() ![]() |
