Assay Details
Gene Symbol: LOC5569526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652790.3 | XP_001652840.2 | 3 - 4 | 745 | 74 | |
XM_021848333.1 | XP_021704025.1 | 2 - 3 | 953 | 74 | ||
XM_021848336.1 | XP_021704028.1 | 3 - 4 | 597 | 74 | ||
XM_021848337.1 | XP_021704029.1 | 3 - 4 | 568 | 74 | ||
GenBank mRNA | AY748242.1 | - | 3 - 4 | 538 | 74 | |
AY748243.1 | - | 2 - 3 | 533 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021848333.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
