Assay Details
Gene Symbol: LOC5569526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652790.3 | XP_001652840.2 | 8 - 9 | 1769 | 80 | |
XM_021848335.1 | XP_021704027.1 | 6 - 7 | 1332 | 80 | ||
XM_021848336.1 | XP_021704028.1 | 8 - 9 | 1621 | 80 | ||
XM_021848337.1 | XP_021704029.1 | 8 - 9 | 1592 | 80 | ||
GenBank mRNA | AY748242.1 | - | 8 - 9 | 1562 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
