Assay Details
Gene Symbol: LOC5571386 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659617.3 | XP_001659667.2 | 2 - 3 | 631 | 79 | |
GenBank mRNA | AY819711.1 | - | 2 - 3 | 546 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
