Assay Details
Gene Symbol: LOC5579996 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840596.1 | XP_021696288.1 | 5 - 6 | 1967 | 77 | |
XM_021840597.1 | XP_021696289.1 | 5 - 6 | 2211 | 77 | ||
XM_021840598.1 | XP_021696290.1 | 6 - 7 | 2277 | 77 | ||
GenBank mRNA | AY902310.1 | - | 4 - 5 | 607 | 77 | |
AY955097.1 | - | 5 - 6 | 1120 | 77 |
