Assay Details
Gene Symbol: LOC5579996 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840596.1 | XP_021696288.1 | 6 - 7 | 2211 | 69 | |
XM_021840597.1 | XP_021696289.1 | 6 - 7 | 2455 | 69 | ||
XM_021840598.1 | XP_021696290.1 | 7 - 8 | 2521 | 69 | ||
GenBank mRNA | AY902310.1 | - | 5 - 6 | 851 | 69 | |
AY955097.1 | - | 6 - 7 | 1364 | 69 |

More Information
Set Membership: |
![]() ![]() |
