Assay Details
Gene Symbol: LOC5579996 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840596.1 | XP_021696288.1 | 9 - 10 | 3644 | 57 | |
XM_021840597.1 | XP_021696289.1 | 9 - 10 | 3888 | 57 | ||
XM_021840598.1 | XP_021696290.1 | 10 - 11 | 3954 | 57 | ||
GenBank mRNA | AY902310.1 | - | 8 - 9 | 2284 | 57 | |
AY955097.1 | - | 9 - 10 | 2797 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
