Assay Details
Gene Symbol: LOC5570907 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653334.2 | XP_001653384.1 | 2 - 3 | 368 | 94 | |
XM_021844938.1 | XP_021700630.1 | 4 - 5 | 597 | 94 | ||
XM_021844939.1 | XP_021700631.1 | 2 - 3 | 395 | 94 | ||
XM_021844940.1 | XP_021700632.1 | 2 - 3 | 399 | 94 | ||
GenBank mRNA | AY944756.1 | - | 1 - 2 | 167 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
