Assay Details
Gene Symbol: LOC5567962 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847385.1 | XP_021703077.1 | 4 - 5 | 3035 | 139 | |
XM_021847386.1 | XP_021703078.1 | 4 - 5 | 3261 | 139 | ||
XM_021847387.1 | XP_021703079.1 | 4 - 5 | 3257 | 139 | ||
XM_021847388.1 | XP_021703080.1 | 4 - 5 | 3261 | 139 | ||
XM_021847389.1 | XP_021703081.1 | 4 - 5 | 3248 | 139 | ||
XM_021847390.1 | XP_021703082.1 | 4 - 5 | 3255 | 139 | ||
XM_021847391.1 | XP_021703083.1 | 4 - 5 | 3255 | 139 | ||
XM_021847392.1 | XP_021703084.1 | 5 - 6 | 3204 | 139 | ||
XM_021847393.1 | XP_021703085.1 | 4 - 5 | 3255 | 139 | ||
XM_021847394.1 | XP_021703086.1 | 4 - 5 | 3255 | 139 | ||
XM_021847395.1 | XP_021703087.1 | 5 - 6 | 3189 | 139 | ||
XM_021847396.1 | XP_021703088.1 | 5 - 6 | 3204 | 139 | ||
XM_021847397.1 | XP_021703089.1 | 4 - 5 | 3256 | 139 | ||
GenBank mRNA | AY971811.1 | - | 1 - 2 | 1756 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() |
