Assay Details
Gene Symbol: LOC5565426 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649754.2 | XP_001649804.1 | 1 - 2 | 886 | 63 | |
GenBank mRNA | CR937211.1 | - | 1 - 2 | 44 | 63 | |
CR937237.1 | - | 1 - 2 | 44 | 63 | ||
CR937259.1 | - | 1 - 2 | 44 | 63 | ||
CR937262.1 | - | 1 - 2 | 44 | 63 | ||
CR937725.1 | - | 1 - 2 | 44 | 63 | ||
CR937872.1 | - | 1 - 2 | 43 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001649754.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
