Assay Details
Gene Symbol: LOC5575604 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001662036.3 | XP_001662086.2 | 1 - 2 | 411 | 94 | |
XM_021838085.1 | XP_021693777.1 | 2 - 3 | 689 | 94 | ||
GenBank mRNA | CR937481.1 | - | 1 - 2 | 416 | 94 |
