Assay Details
Gene Symbol: LOC5572775 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654127.2 | XP_001654177.1 | 1 - 2 | 374 | 79 | |
GenBank mRNA | CR937578.1 | - | 1 - 2 | 82 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001654127.2 transcripts.
Set Membership: |
![]() ![]() |
