Assay Details
Gene Symbol: LOC5572775 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654127.2 | XP_001654177.1 | 2 - 3 | 529 | 121 | |
GenBank mRNA | CR937578.1 | - | 2 - 3 | 237 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
