Assay Details
Gene Symbol: LOC5564327 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001648604.2 | XP_001648654.2 | 3 | 1932 | 69 | |
GenBank mRNA | CR937882.1 | - | 1 | 820 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
