Assay Details
Gene Symbol: LOC5566228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001650566.2 | XP_001650616.1 | 2 - 3 | 370 | 98 | |
XM_001650567.2 | XP_001650617.1 | 2 - 3 | 335 | 98 | ||
GenBank mRNA | CR938003.1 | - | 1 - 2 | 103 | 98 | |
DQ440270.1 | - | 1 - 2 | 232 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
