Assay Details
Gene Symbol: LOC5574515 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852477.1 | XP_021708169.1 | 1 - 2 | 154 | 60 | |
GenBank mRNA | CR938145.1 | - | 1 - 2 | 53 | 60 | |
CR938607.1 | - | 1 - 2 | 38 | 60 | ||
CR938643.1 | - | 1 - 2 | 34 | 60 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021852477.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
