Assay Details
Gene Symbol: LOC5577613 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656557.2 | XP_001656607.2 | 2 - 3 | 556 | 81 | |
GenBank mRNA | CR938427.1 | - | 2 - 3 | 442 | 81 | |
CR938690.1 | - | 2 - 3 | 450 | 81 |
