Assay Details
Gene Symbol: LOC5577613 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656557.2 | XP_001656607.2 | 3 - 4 | 799 | 66 | |
GenBank mRNA | CR938427.1 | - | 3 - 4 | 685 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
