Assay Details
Gene Symbol: LOC5567888 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651873.2 | XP_001651923.1 | 1 - 2 | 292 | 70 | |
GenBank mRNA | CR938479.1 | - | 1 - 2 | 274 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
