Assay Details
Gene Symbol: LOC5573257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654418.2 | XP_001654468.1 | 4 - 5 | 1345 | 87 | |
XM_021847231.1 | XP_021702923.1 | 4 - 5 | 1277 | 87 | ||
GenBank mRNA | CR938489.1 | - | 1 - 2 | 137 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
