Assay Details
Gene Symbol: LOC5573257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654418.2 | XP_001654468.1 | 5 - 6 | 1481 | 78 | |
XM_021847231.1 | XP_021702923.1 | 5 - 6 | 1413 | 78 | ||
GenBank mRNA | CR938489.1 | - | 2 - 3 | 273 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
