Assay Details
Gene Symbol: LOC5573257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654418.2 | XP_001654468.1 | 6 - 7 | 1646 | 86 | |
XM_021847231.1 | XP_021702923.1 | 6 - 7 | 1578 | 86 | ||
GenBank mRNA | CR938489.1 | - | 3 - 4 | 438 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
