Assay Details
Gene Symbol: LOC5567158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651539.2 | XP_001651589.1 | 1 - 2 | 276 | 93 | |
GenBank mRNA | DQ440102.1 | - | 1 - 2 | 65 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
