Assay Details
Gene Symbol: LOC5567158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651539.2 | XP_001651589.1 | 2 - 3 | 951 | 91 | |
GenBank mRNA | DQ440102.1 | - | 2 - 3 | 740 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
