Assay Details
Gene Symbol: LOC5573216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654381.2 | XP_001654431.1 | 4 | 1908 | 95 | |
GenBank mRNA | DQ440118.1 | - | 1 | 917 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
