Assay Details
Gene Symbol: LOC5574137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661156.2 | XP_001661206.1 | 2 - 3 | 539 | 69 | |
XM_021851631.1 | XP_021707323.1 | 2 - 3 | 538 | 69 | ||
XM_021851632.1 | XP_021707324.1 | 2 - 3 | 379 | 69 | ||
GenBank mRNA | DQ440127.1 | - | 1 - 2 | 281 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
