Assay Details
Gene Symbol: LOC5572447 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001660341.2 | XP_001660391.1 | 1 - 2 | 176 | 81 | |
GenBank mRNA | DQ440158.1 | - | 1 - 2 | 67 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
