Assay Details
Gene Symbol: LOC5572447 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001660341.2 | XP_001660391.1 | 2 - 3 | 306 | 77 | |
GenBank mRNA | DQ440158.1 | - | 2 - 3 | 197 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
