Assay Details
Gene Symbol: LOC5575622 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001662060.2 | XP_001662110.1 | 1 - 2 | 707 | 105 | |
XM_021844005.1 | XP_021699697.1 | 2 - 3 | 631 | 105 | ||
XM_021844006.1 | XP_021699698.1 | 2 - 3 | 509 | 105 | ||
GenBank mRNA | DQ440159.1 | - | 1 - 2 | 127 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
