Assay Details
Gene Symbol: LOC5568313 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652140.2 | XP_001652190.1 | 1 - 2 | 358 | 72 | |
GenBank mRNA | DQ440162.1 | - | 1 - 2 | 226 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
