Assay Details
Gene Symbol: LOC5570147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658898.2 | XP_001658948.1 | 1 - 2 | 65 | ||
GenBank mRNA | DQ440164.1 | - | 1 - 2 | 102 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
