Assay Details
Gene Symbol: LOC5571507 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848418.1 | XP_021704110.1 | 2 - 3 | 2992 | 77 | |
XM_021848419.1 | XP_021704111.1 | 2 - 3 | 2994 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
