Assay Details
Gene Symbol: LOC5571507 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848418.1 | XP_021704110.1 | 1 - 2 | 684 | 87 | |
XM_021848419.1 | XP_021704111.1 | 1 - 2 | 686 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
