Assay Details
Gene Symbol: LOC5572484 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849710.1 | XP_021705402.1 | 17 - 18 | 7844 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5572484 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849710.1 | XP_021705402.1 | 17 - 18 | 7844 | 106 |
Set Membership: |
![]() ![]() ![]() |