Assay Details
Gene Symbol: LOC5576852 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656230.2 | XP_001656280.2 | 3 - 4 | 704 | 91 | |
XM_021841836.1 | XP_021697528.1 | 3 - 4 | 479 | 91 | ||
XM_021841837.1 | XP_021697529.1 | 2 - 3 | 236 | 91 | ||
XM_021841838.1 | XP_021697530.1 | 2 - 3 | 434 | 91 |
