Assay Details
Gene Symbol: LOC5576852 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656230.2 | XP_001656280.2 | 5 - 6 | 962 | 57 | |
XM_021841836.1 | XP_021697528.1 | 5 - 6 | 737 | 57 | ||
XM_021841837.1 | XP_021697529.1 | 4 - 5 | 494 | 57 | ||
XM_021841838.1 | XP_021697530.1 | 4 - 5 | 692 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
