Assay Details
Gene Symbol: LOC5576852 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656230.2 | XP_001656280.2 | 7 - 8 | 1272 | 72 | |
XM_021841836.1 | XP_021697528.1 | 7 - 8 | 1047 | 72 | ||
XM_021841837.1 | XP_021697529.1 | 6 - 7 | 804 | 72 | ||
XM_021841838.1 | XP_021697530.1 | 6 - 7 | 1002 | 72 |
