Assay Details
Gene Symbol: LOC5568103 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651988.2 | XP_001652038.1 | 3 - 4 | 448 | 73 | |
XM_011495041.2 | XP_011493343.1 | 2 - 3 | 318 | 73 | ||
XM_021840936.1 | XP_021696628.1 | 3 - 4 | 704 | 73 | ||
XM_021840937.1 | XP_021696629.1 | 3 - 4 | 480 | 73 | ||
XM_021840938.1 | XP_021696630.1 | 3 - 4 | 207 | 73 | ||
GenBank mRNA | DQ440336.1 | - | 1 - 2 | 84 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
