Assay Details
Gene Symbol: LOC5567565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001657506.2 | XP_001657556.2 | 1 - 2 | 163 | 148 | |
GenBank mRNA | AY432885.1 | - | 1 - 2 | 82 | 148 | |
CR938042.1 | - | 1 - 2 | 82 | 148 | ||
M95187.1 | - | 1 - 2 | 71 | 148 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001657506.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
