Assay Details
Gene Symbol: LOC5574421 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661388.2 | XP_001661438.1 | 3 - 4 | 564 | 79 | |
GenBank mRNA | AY432886.1 | - | 3 - 4 | 472 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
