Assay Details
Gene Symbol: LOC5575813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001662189.3 | XP_001662239.2 | 2 - 3 | 283 | 64 | |
XM_021849765.1 | XP_021705457.1 | 2 - 3 | 352 | 64 | ||
XM_021849766.1 | XP_021705458.1 | 2 - 3 | 479 | 64 | ||
GenBank mRNA | AY432890.1 | - | 2 - 3 | 347 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
