Assay Details
Gene Symbol: LOC5579027 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001664158.2 | XP_001664208.1 | 7 | 2777 | 90 | |
XM_021853258.1 | XP_021708950.1 | 7 | 2799 | 90 | ||
XM_021853259.1 | XP_021708951.1 | 7 | 2573 | 90 | ||
XM_021853260.1 | XP_021708952.1 | 7 | 2505 | 90 | ||
GenBank mRNA | AY432916.1 | - | 1 | 214 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
