Assay Details
Gene Symbol: LOC5571395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653597.2 | XP_001653647.2 | 1 - 2 | 223 | 93 | |
GenBank mRNA | AY432942.1 | - | 1 - 2 | 218 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001653597.2 transcripts.
Set Membership: |
![]() ![]() |
