Assay Details
Gene Symbol: LOC5571395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653597.2 | XP_001653647.2 | 2 - 3 | 393 | 87 | |
GenBank mRNA | AY432942.1 | - | 2 - 3 | 388 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
