Assay Details
Gene Symbol: LOC5576642 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845927.1 | XP_021701619.1 | 2 - 3 | 442 | 83 | |
GenBank mRNA | DQ440382.1 | - | 1 - 2 | 124 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
