Assay Details
Gene Symbol: LOC5576642 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845927.1 | XP_021701619.1 | 3 - 4 | 572 | 101 | |
GenBank mRNA | DQ440382.1 | - | 2 - 3 | 254 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
